Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122892.1 | NP_001116364.1 | 11 - 12 | 1703 | 71 | |
NM_001122893.1 | NP_001116365.1 | 12 - 13 | 1782 | 71 | ||
NM_008054.2 | NP_032080.2 | 12 - 13 | 1773 | 71 | ||
GenBank mRNA | AK156584.1 | - | 12 - 13 | 1784 | 71 | |
BC010554.1 | - | 11 - 12 | 1384 | 71 | ||
BC032149.1 | - | 11 - 12 | 1418 | 3602546 | 71 | |
BC092217.1 | - | 11 - 12 | 1699 | 6403711 | 71 | |
M27266.1 | - | 13 - 14 | 1485 | 71 | ||
U70324.1 | - | 12 - 13 | 1486 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |