Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmym6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC140958.1 | - | 8 - 9 | 1232 | 9055570 | 75 |
BC144807.1 | - | 8 - 9 | 1239 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |