Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Moxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021509.5 | NP_067484.2 | 1 - 2 | 353 | 82 | |
GenBank mRNA | AB041606.1 | - | 1 - 2 | 288 | 82 | |
AK014340.1 | - | 1 - 2 | 354 | 82 | ||
AK030274.1 | - | 1 - 2 | 294 | 82 | ||
AK081586.1 | - | 1 - 2 | 319 | 82 | ||
AK162030.1 | - | 1 - 2 | 295 | 82 | ||
BC037684.1 | - | 1 - 2 | 291 | 82 | ||
BC057652.1 | - | 1 - 2 | 267 | 4017199 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |