Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3mbtl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172787.2 | NP_766375.1 | 2 - 3 | 195 | 88 | |
GenBank mRNA | AK031398.1 | - | 2 - 3 | 181 | 88 | |
AK045667.1 | - | 2 - 3 | 196 | 88 | ||
AK049597.1 | - | 2 - 3 | 230 | 88 | ||
BC082309.1 | - | 2 - 3 | 140 | 30356672 | 88 | |
BC085192.1 | - | 2 - 3 | 375 | 30694487 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |