Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023665.3 | NP_076154.3 | 1 - 2 | 147 | 61 | |
GenBank mRNA | AB041555.1 | - | 1 - 2 | 127 | 61 | |
AK003799.1 | - | 1 - 2 | 148 | 61 | ||
AK012363.1 | - | 1 - 2 | 148 | 61 | ||
AK028145.1 | - | 1 - 2 | 149 | 61 | ||
AK029529.1 | - | 1 - 2 | 147 | 61 | ||
AK150024.1 | - | 1 - 2 | 148 | 61 | ||
AK150665.1 | - | 1 - 2 | 127 | 61 | ||
AK152736.1 | - | 1 - 2 | 127 | 61 | ||
AK160993.1 | - | 1 - 2 | 148 | 61 | ||
AK169598.1 | - | 1 - 2 | 149 | 61 | ||
AK169760.1 | - | 1 - 2 | 148 | 61 | ||
BC002142.1 | - | 1 - 2 | 136 | 61 | ||
BC032060.1 | - | 1 - 2 | 144 | 61 | ||
BC043057.1 | - | 1 - 2 | 132 | 6419597 | 61 | |
BC045256.1 | - | 1 - 2 | 136 | 61 | ||
BC046231.1 | - | 1 - 2 | 49 | 6813388 | 61 | |
BC051556.1 | - | 1 - 2 | 142 | 61 | ||
BC054728.1 | - | 1 - 2 | 120 | 6419723 | 61 | |
BC056986.1 | - | 1 - 2 | 81 | 6816649 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |