Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130477.1 | NP_001123949.1 | 1 - 2 | 118 | 74 | |
NM_016799.3 | NP_058079.2 | 1 - 2 | 118 | 74 | ||
GenBank mRNA | AF062655.1 | - | 1 - 2 | 44 | 74 | |
AK008284.1 | - | 1 - 2 | 46 | 74 | ||
AK028412.1 | - | 1 - 2 | 50 | 74 | ||
AK169762.1 | - | 44 | 74 | |||
BC079542.1 | - | 1 - 2 | 38 | 74 | ||
BC094322.1 | - | 1 - 2 | 33 | 6401250 | 74 | |
BU514596.1 | - | 1 - 2 | 118 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |