Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282118.1 | NP_001269047.1 | 10 - 11 | 1742 | 87 | |
NM_001282119.1 | NP_001269048.1 | 10 - 11 | 1664 | 87 | ||
NM_001282120.1 | NP_001269049.1 | 10 - 11 | 2106 | 87 | ||
NM_133767.3 | NP_598528.2 | 10 - 11 | 2020 | 87 | ||
GenBank mRNA | AK054313.1 | - | 1 - 2 | 894 | 87 | |
AK136344.1 | - | 10 - 11 | 1704 | 87 | ||
AK146961.1 | - | 10 - 11 | 1960 | 87 | ||
BC016590.1 | - | 10 - 11 | 1596 | 3585359 | 87 | |
BC066819.1 | - | 10 - 11 | 2000 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |