Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146016.2 | NP_666128.2 | 15 - 16 | 2903 | 63 | |
GenBank mRNA | AK049034.1 | - | 15 - 16 | 2815 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |