Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177629.1 | NP_001171100.1 | 2 - 3 | 144 | 93 | |
NM_010345.4 | NP_034475.2 | 2 - 3 | 304 | 93 | ||
GenBank mRNA | AB106541.1 | - | 2 - 3 | 562 | 93 | |
AF022072.1 | - | 2 - 3 | 278 | 93 | ||
AK012646.1 | - | 2 - 3 | 284 | 93 | ||
AK030727.1 | - | 2 - 3 | 239 | 93 | ||
AK032927.1 | - | 2 - 3 | 285 | 93 | ||
AK163841.1 | - | 2 - 3 | 285 | 93 | ||
BC016111.1 | - | 2 - 3 | 144 | 4481345 | 93 | |
BC053842.1 | - | 2 - 3 | 271 | 93 | ||
BY122902.1 | - | 2 - 3 | 126 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |