Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172546.2 | NP_766134.1 | 10 - 11 | 1620 | 114 | |
GenBank mRNA | AK033015.1 | - | 10 - 11 | 1621 | 114 | |
AK149951.1 | - | 9 - 10 | 919 | 114 | ||
AY151137.1 | - | 10 - 11 | 1582 | 114 | ||
BC024086.1 | - | 9 - 10 | 968 | 114 | ||
BC031194.1 | - | 3 - 4 | 727 | 5059479 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |