Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172546.2 | NP_766134.1 | 9 - 10 | 1502 | 90 | |
GenBank mRNA | AK033015.1 | - | 9 - 10 | 1503 | 90 | |
AK149951.1 | - | 8 - 9 | 801 | 90 | ||
AY151137.1 | - | 9 - 10 | 1464 | 90 | ||
BC024086.1 | - | 8 - 9 | 850 | 90 | ||
BC031194.1 | - | 2 - 3 | 609 | 5059479 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |