Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009260.2 | NP_033286.2 | 26 - 27 | 6208 | 114 | |
NM_175836.2 | NP_787030.2 | 26 - 27 | 5868 | 114 | ||
GenBank mRNA | AF017112.1 | - | 26 - 27 | 6115 | 114 | |
AK051630.1 | - | 1 - 2 | 226 | 114 | ||
AK161119.1 | - | 5 - 6 | 991 | 114 | ||
AK220456.1 | - | 27 - 28 | 6070 | 114 | ||
M74773.1 | - | 26 - 27 | 6188 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |