Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009260.2 | NP_033286.2 | 5 - 6 | 1029 | 112 | |
NM_175836.2 | NP_787030.2 | 5 - 6 | 689 | 112 | ||
GenBank mRNA | AF017112.1 | - | 5 - 6 | 936 | 112 | |
AK157727.1 | - | 5 - 6 | 1019 | 112 | ||
AK169544.1 | - | 5 - 6 | 1029 | 112 | ||
AK220456.1 | - | 6 - 7 | 891 | 112 | ||
M74773.1 | - | 5 - 6 | 1009 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |