Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290753.1 | NP_001277682.1 | 14 - 15 | 2881 | 86 | |
NM_010142.3 | NP_034272.1 | 14 - 15 | 2878 | 86 | ||
GenBank mRNA | BC043088.1 | - | 14 - 15 | 2802 | 6418088 | 86 |
BC062924.1 | - | 14 - 15 | 2826 | 86 | ||
L25890.1 | - | 15 - 16 | 2720 | 86 | ||
X76011.1 | - | 8 - 9 | 1179 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |