Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175554.4 | NP_780763.3 | 1 - 2 | 160 | 91 | |
GenBank mRNA | AK045076.1 | - | 1 - 2 | 156 | 91 | |
AK051966.1 | - | 1 - 2 | 135 | 91 | ||
AK053847.1 | - | 1 - 2 | 174 | 91 | ||
AK135394.1 | - | 1 - 2 | 154 | 91 | ||
BC050848.1 | - | 1 - 2 | 115 | 6507692 | 91 | |
CF535627.1 | - | 1 - 2 | 160 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |