Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rlf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081013.1 | NP_001074482.1 | 3 - 4 | 659 | 134 | |
GenBank mRNA | AK132452.1 | - | 2 - 3 | 449 | 134 | |
AK162649.1 | - | 5 - 6 | 774 | 134 | ||
AK163144.1 | - | 2 - 3 | 282 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |