Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600005554.mm.1 |
TC0600000724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600005554.mm.1 |
NM_001163507.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005554.mm.1 |
NM_001163507.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005554.mm.1 |
NM_027613.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005554.mm.1 |
NM_027613.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005556.mm.1 |
TC0600000724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600005556.mm.1 |
NM_001163507.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005556.mm.1 |
NM_001163507.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005556.mm.1 |
NM_027613.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005556.mm.1 |
NM_027613.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005552.mm.1 |
TC0600000724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600005552.mm.1 |
NM_001163507.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005552.mm.1 |
NM_001163507.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005552.mm.1 |
NM_027613.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005552.mm.1 |
NM_027613.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005553.mm.1 |
TC0600000724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600005553.mm.1 |
NM_001163507.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005553.mm.1 |
NM_001163507.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005553.mm.1 |
NM_027613.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005553.mm.1 |
NM_027613.1 |
Refseq |