Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027572.1 | NP_081848.1 | 2 - 3 | 647 | 108 | |
GenBank mRNA | BC100473.1 | - | 2 - 3 | 647 | 30918267 | 108 |
BC138326.1 | - | 2 - 3 | 658 | 8861346 | 108 | |
BC138327.1 | - | 2 - 3 | 658 | 8861347 | 108 | |
DQ164804.2 | - | 2 - 3 | 673 | 108 | ||
DQ164805.1 | - | 2 - 3 | 634 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |