Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB021291.1 | - | 5 - 6 | 647 | 108 | |
AF038652.1 | - | 4 - 5 | 406 | 108 | ||
AK035993.1 | - | 5 - 6 | 679 | 108 | ||
AK047067.1 | - | 5 - 6 | 663 | 108 | ||
AK143196.1 | - | 5 - 6 | 691 | 109 | ||
AK158292.1 | - | 5 - 6 | 613 | 108 | ||
BC090625.1 | - | 5 - 6 | 649 | 108 | ||
BC094567.1 | - | 5 - 6 | 674 | 6834138 | 108 | |
BC139232.1 | - | 5 - 6 | 621 | 8862254 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |