Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF040094.2 | - | 16 - 17 | 2147 | 99 | |
AK004601.1 | - | 17 - 18 | 2154 | 99 | ||
AK004722.2 | - | 16 - 17 | 2049 | 99 | ||
AY007563.1 | - | 23 - 24 | 2932 | 99 | ||
BC028864.1 | - | 22 - 23 | 2865 | 4457437 | 99 | |
BG802289.1 | - | 1 - 2 | 140 | 99 | ||
DV654857.1 | - | 5 - 6 | 717 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |