Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017383.3 | NP_059079.2 | 8 - 9 | 1264 | 124 | |
GenBank mRNA | AB032602.1 | - | 7 - 8 | 950 | 124 | |
AK052972.1 | - | 9 - 10 | 1157 | 124 | ||
BC076594.1 | - | 8 - 9 | 1264 | 30649661 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |