Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011630.3 | NP_035760.1 | 3 - 4 | 476 | 77 | |
GenBank mRNA | AK157305.1 | - | 3 - 4 | 450 | 77 | |
BC103683.1 | - | 1 - 2 | 95 | 40047943 | 77 | |
BC103684.1 | - | 1 - 2 | 95 | 40047942 | 77 | |
BC103686.1 | - | 1 - 2 | 95 | 40047941 | 77 | |
S75970.1 | - | 3 - 4 | 591 | 77 | ||
U11688.1 | - | 2 - 3 | 138 | 77 | ||
U32939.1 | - | 1 - 2 | 109 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons |