Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081111.2 | NP_001074580.1 | 15 - 16 | 3109 | 80 | |
GenBank mRNA | BC150931.1 | - | 15 - 16 | 3120 | 9087842 | 80 |
BQ561256.1 | - | 2 - 3 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |