Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600021696.mm.1 |
TC0600002797.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021696.mm.1 |
NM_001081111.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021696.mm.1 |
NM_001081111.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021695.mm.1 |
TC0600002797.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021695.mm.1 |
NM_001081111.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021695.mm.1 |
NM_001081111.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021697.mm.1 |
TC0600002797.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021697.mm.1 |
NM_001081111.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021697.mm.1 |
NM_001081111.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021698.mm.1 |
TC0600002797.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021698.mm.1 |
NM_001081111.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021698.mm.1 |
NM_001081111.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021694.mm.1 |
TC0600002797.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021694.mm.1 |
NM_001081111.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021694.mm.1 |
NM_001081111.2 |
Refseq |