Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uroc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144940.2 | NP_659189.2 | 10 - 11 | 999 | 64 | |
GenBank mRNA | AK149464.1 | - | 10 - 11 | 1102 | 64 | |
BC022133.1 | - | 10 - 11 | 5098655 | 64 | ||
BC033269.1 | - | 7 - 8 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |