Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqsec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134383.1 | NP_001127855.1 | 13 - 14 | 3104 | 87 | |
GenBank mRNA | AK220355.1 | - | 11 - 12 | 1555 | 87 | |
BC026481.1 | - | 11 - 12 | 1608 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |