Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144918.2 | NP_659167.2 | 3 - 4 | 375 | 71 | |
GenBank mRNA | AK158267.1 | - | 3 - 4 | 356 | 71 | |
AK162926.1 | - | 3 - 4 | 299 | 71 | ||
BC016525.1 | - | 3 - 4 | 362 | 3601356 | 71 | |
BU847995.1 | - | 3 - 4 | 378 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |