Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144918.2 | NP_659167.2 | 4 - 5 | 489 | 84 | |
GenBank mRNA | AK158267.1 | - | 4 - 5 | 470 | 84 | |
AK162926.1 | - | 4 - 5 | 413 | 84 | ||
BC016525.1 | - | 4 - 5 | 476 | 3601356 | 84 | |
BU847995.1 | - | 4 - 5 | 492 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |