Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cav3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007617.3 | NP_031643.1 | 1 - 2 | 422 | 64 | |
GenBank mRNA | BC024383.1 | - | 1 - 2 | 432 | 4988598 | 64 |
CJ057698.1 | - | 1 - 2 | 199 | 64 | ||
U36579.1 | - | 1 - 2 | 236 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |