Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK154976.1 | - | 1 - 2 | 197 | 69 | |
AK165647.1 | - | 1 - 2 | 165 | 69 | ||
BC050908.1 | - | 1 - 2 | 180 | 6416955 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |