Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700031653.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031653.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031653.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031651.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031651.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031651.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031649.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031649.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031649.mm.1 |
TC0700003763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031655.mm.1 |
NM_001177412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031655.mm.1 |
NM_001177412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031655.mm.1 |
TC0700003763.mm.1 |
MTA TC |