Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138592.4 | NP_613058.1 | 1 - 2 | 290 | 78 | |
GenBank mRNA | AK010443.1 | - | 1 - 2 | 300 | 78 | |
AK018595.1 | - | 1 - 2 | 291 | 78 | ||
AK033105.1 | - | 1 - 2 | 287 | 78 | ||
AK045062.1 | - | 1 - 2 | 287 | 78 | ||
AK088354.1 | - | 1 - 2 | 302 | 78 | ||
AK165625.1 | - | 1 - 2 | 271 | 78 | ||
AK167668.1 | - | 1 - 2 | 278 | 78 | ||
AK168003.1 | - | 1 - 2 | 297 | 78 | ||
AK201881.1 | - | 1 - 2 | 57 | 78 | ||
BC026983.1 | - | 1 - 2 | 265 | 5365862 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |