Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016886.1 | - | 9 - 10 | 619 | 80 | |
BC100525.1 | - | 9 - 10 | 576 | 30920200 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |