Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011708.4 | NP_035838.3 | 23 - 24 | 3359 | 75 | |
GenBank mRNA | AF539800.2 | - | 22 - 23 | 3107 | 75 | |
AK053806.1 | - | 1 - 2 | 1348 | 75 | ||
AK157296.1 | - | 23 - 24 | 3361 | 75 | ||
AK170201.1 | - | 10 - 11 | 1719 | 75 | ||
AY208897.1 | - | 23 - 24 | 3167 | 75 | ||
DQ355287.1 | - | 23 - 24 | 3344 | 75 | ||
DQ355288.1 | - | 23 - 24 | 3344 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |