Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011708.4 | NP_035838.3 | 46 - 47 | 8021 | 62 | |
GenBank mRNA | AF539800.2 | - | 45 - 46 | 7769 | 62 | |
AK078523.1 | - | 13 - 14 | 1881 | 62 | ||
AY208897.1 | - | 46 - 47 | 7829 | 62 | ||
DQ355287.1 | - | 46 - 47 | 8006 | 62 | ||
DQ355288.1 | - | 46 - 47 | 8006 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |