Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198604.2 | NP_941006.2 | 3 - 4 | 588 | 144 | |
GenBank mRNA | AK153619.1 | - | 3 - 4 | 878 | 144 | |
BC026778.1 | - | 3 - 4 | 591 | 5133546 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |