Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198604.2 | NP_941006.2 | 7 - 8 | 73 | ||
GenBank mRNA | AK153619.1 | - | 7 - 8 | 73 | ||
BC026778.1 | - | 7 - 8 | 1053 | 5133546 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |