Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scnn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011324.2 | NP_035454.2 | 9 - 10 | 2120 | 72 | |
GenBank mRNA | AF112185.1 | - | 9 - 10 | 1607 | 72 | |
AK140389.1 | - | 9 - 10 | 2121 | 72 | ||
BC040814.1 | - | 9 - 10 | 1750 | 72 | ||
BC049952.1 | - | 7 - 8 | 711 | 72 | ||
BC133688.1 | - | 9 - 10 | 1601 | 40111601 | 72 | |
BC133689.1 | - | 9 - 10 | 1601 | 40111602 | 72 | |
U52006.1 | - | 3 - 4 | 244 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |