Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138677.2 | NP_619618.1 | 1 - 2 | 591 | 108 | |
GenBank mRNA | AB042828.1 | - | 1 - 2 | 580 | 108 | |
AK030943.1 | - | 1 - 2 | 592 | 108 | ||
AK037889.1 | - | 1 - 2 | 154 | 108 | ||
AK082920.1 | - | 1 - 2 | 544 | 108 | ||
AK129090.1 | - | 1 - 2 | 558 | 108 | ||
AK151668.1 | - | 1 - 2 | 566 | 108 | ||
AK152724.1 | - | 1 - 2 | 567 | 108 | ||
BC023237.1 | - | 1 - 2 | 539 | 2654113 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |