Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008894.2 | NP_032920.2 | 7 - 8 | 979 | 65 | |
GenBank mRNA | AK013901.1 | - | 7 - 8 | 945 | 65 | |
BC003342.1 | - | 7 - 8 | 945 | 3257610 | 65 | |
CB205181.1 | - | 4 - 5 | 515 | 65 | ||
Z72486.1 | - | 7 - 8 | 940 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |