Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207210.2 | NP_997093.2 | 2 - 3 | 186 | 66 | |
GenBank mRNA | AK077117.1 | - | 2 - 3 | 187 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mus musculus Probe spans exons |