Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207210.2 | NP_997093.2 | 9 - 10 | 1097 | 105 | |
GenBank mRNA | AK077117.1 | - | 9 - 10 | 1098 | 105 | |
BC052324.1 | - | 8 - 9 | 1151 | 6315342 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |