Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007544.3 | NP_031570.2 | 2 - 3 | 155 | 91 | |
GenBank mRNA | AK045731.1 | - | 2 - 3 | 170 | 91 | |
AK051076.1 | - | 171 | 91 | |||
AK052356.1 | - | 2 - 3 | 160 | 91 | ||
AK077657.1 | - | 2 - 3 | 138 | 91 | ||
AK161235.1 | - | 2 - 3 | 177 | 91 | ||
BC002031.1 | - | 2 - 3 | 155 | 3486537 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus 1700 Probe spans exons |