Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cecr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128151.1 | NP_001121623.1 | 10 - 11 | 1401 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |