Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145997.2 | NP_666109.2 | 9 - 10 | 1658 | 109 | |
GenBank mRNA | AK144877.1 | - | 9 - 10 | 1629 | 109 | |
AK159079.1 | - | 9 - 10 | 1635 | 109 | ||
AK166055.1 | - | 9 - 10 | 1639 | 109 | ||
BC010717.1 | - | 9 - 10 | 1299 | 109 | ||
BC011506.3 | - | 9 - 10 | 1592 | 109 | ||
BC080691.1 | - | 9 - 10 | 1344 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |