Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204223.1 | NP_001191152.1 | 2 - 3 | 192 | 68 | |
NM_027709.2 | NP_081985.1 | 1 - 2 | 106 | 68 | ||
GenBank mRNA | AK016908.1 | - | 1 - 2 | 107 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |