Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081437.1 | NP_001074906.1 | 15 - 16 | 3199 | 113 | |
NM_007992.2 | NP_032018.2 | 16 - 17 | 3340 | 113 | ||
GenBank mRNA | AK159361.1 | - | 15 - 16 | 3511 | 113 | |
AK161522.1 | - | 15 - 16 | 3142 | 113 | ||
AK168688.1 | - | 15 - 16 | 3137 | 113 | ||
BC005443.1 | - | 15 - 16 | 3179 | 3490759 | 113 | |
X75285.1 | - | 16 - 17 | 3240 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |