Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011585.4 | NP_035715.1 | 5 - 6 | 586 | 67 | |
GenBank mRNA | AK088486.1 | - | 5 - 6 | 522 | 67 | |
AK144222.1 | - | 5 - 6 | 539 | 67 | ||
AK185472.1 | - | 3 - 4 | 104 | 67 | ||
U00689.1 | - | 5 - 6 | 527 | 67 | ||
U55862.1 | - | 5 - 6 | 394 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mus musculus Probe spans exons |