Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Figla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012013.1 | NP_036143.1 | 1 - 2 | 261 | 83 | |
GenBank mRNA | JN700518.1 | - | 1 - 2 | 221 | 83 | |
U91840.1 | - | 1 - 2 | 261 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |