Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133933.4 | NP_598694.3 | 1 - 2 | 323 | 141 | |
GenBank mRNA | AK029923.1 | - | 1 - 2 | 281 | 141 | |
AK132750.1 | - | 1 - 2 | 296 | 141 | ||
AK151899.1 | - | 1 - 2 | 287 | 141 | ||
AK152003.1 | - | 1 - 2 | 296 | 141 | ||
AK153867.1 | - | 1 - 2 | 282 | 141 | ||
BC016080.1 | - | 1 - 2 | 285 | 4489287 | 141 | |
BC085483.1 | - | 1 - 2 | 264 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |